missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEPES Hemisodium Salt, MP Biomedicals™
$203.29 - $203.29
Chemical Identifiers
| CAS | 103404-87-1 |
|---|---|
| Molecular Formula | C8H17N2NaO4S |
| Molecular Weight (g/mol) | 260.28 |
| MDL Number | MFCD00036463 |
| InChI Key | RDZTWEVXRGYCFV-UHFFFAOYSA-M |
| Synonym | hepes sodium salt, hepes hemisodium salt, sodium 2-4-2-hydroxyethyl piperazin-1-yl ethanesulfonate, unii-z9fto91o8a, 1-piperazineethanesulfonic acid, 4-2-hydroxyethyl-, monosodium salt, z9fto91o8a, sodium 4-2-hydroxyethyl piperazin-1-ylethanesulphonate, 4-2-hydroxyethyl piperazine-1-ethanesulfonic acid sodium salt, 4-2-hydroxyethyl piperazine-1-ethanesulfonic acid hemisodium salt |
| PubChem CID | 2724248 |
| ChEBI | CHEBI:46758 |
| IUPAC Name | sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate |
| SMILES | [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15245180
|
MP Biomedicals Inc
0215245180 |
100 g |
Each for $203.29
|
|
|||||
Chemical Identifiers
| 103404-87-1 | |
| 260.28 | |
| RDZTWEVXRGYCFV-UHFFFAOYSA-M | |
| 2724248 | |
| sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate |
| C8H17N2NaO4S | |
| MFCD00036463 | |
| hepes sodium salt, hepes hemisodium salt, sodium 2-4-2-hydroxyethyl piperazin-1-yl ethanesulfonate, unii-z9fto91o8a, 1-piperazineethanesulfonic acid, 4-2-hydroxyethyl-, monosodium salt, z9fto91o8a, sodium 4-2-hydroxyethyl piperazin-1-ylethanesulphonate, 4-2-hydroxyethyl piperazine-1-ethanesulfonic acid sodium salt, 4-2-hydroxyethyl piperazine-1-ethanesulfonic acid hemisodium salt | |
| CHEBI:46758 | |
| [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1 |
Specifications
| 103404-87-1 | |
| 1.005 | |
| MFCD00036463 | |
| hepes sodium salt, hepes hemisodium salt, sodium 2-4-2-hydroxyethyl piperazin-1-yl ethanesulfonate, unii-z9fto91o8a, 1-piperazineethanesulfonic acid, 4-2-hydroxyethyl-, monosodium salt, z9fto91o8a, sodium 4-2-hydroxyethyl piperazin-1-ylethanesulphonate, 4-2-hydroxyethyl piperazine-1-ethanesulfonic acid sodium salt, 4-2-hydroxyethyl piperazine-1-ethanesulfonic acid hemisodium salt | |
| [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1 | |
| 260.28 | |
| CHEBI:46758 | |
| Powder |
| White | |
| C8H17N2NaO4S | |
| 100 g | |
| RDZTWEVXRGYCFV-UHFFFAOYSA-M | |
| sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate | |
| 2724248 | |
| 249.3 | |
| HEPES Hemisodium Salt |
Safety and Handling
Recommended Storage : Store at Room Temperature (15 to 30°C). Store Desiccated.