missing translation for 'onlineSavingsMsg'
Learn More

HEPES, free acid, >98%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0210192690

 View more versions of this product

Catalog No. ICN10192690


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

HEPES
White
C8H18N2O4S
500 g
JKMHFZQWWAIEOD-UHFFFAOYSA-N
sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
23831
238.3
7365-45-9
>98%
MFCD00006158
hepes, 2-4-2-hydroxyethyl piperazin-1-yl ethanesulfonic acid, 4-2-hydroxyethyl-1-piperazineethanesulfonic acid, 1-piperazineethanesulfonic acid, 4-2-hydroxyethyl, monosodium salt, hepes, 4-2-hydroxyethyl piperazine-1-ethanesulfonic acid, unii-rww266ye9i, 4-2-hydroxyethyl-1-piperazineethane sulfonic acid, n-2-hydroxyethylpiperazine-n'-ethanesulfonate
OCCN1CCN(CCS(O)(=O)=O)CC1
238.30
CHEBI:42334
Powder

Chemical Identifiers

7365-45-9
238.30
JKMHFZQWWAIEOD-UHFFFAOYSA-N
23831
OCCN1CCN(CCS(O)(=O)=O)CC1
C8H18N2O4S
MFCD00006158
hepes, 2-4-2-hydroxyethyl piperazin-1-yl ethanesulfonic acid, 4-2-hydroxyethyl-1-piperazineethanesulfonic acid, 1-piperazineethanesulfonic acid, 4-2-hydroxyethyl, monosodium salt, hepes, 4-2-hydroxyethyl piperazine-1-ethanesulfonic acid, unii-rww266ye9i, 4-2-hydroxyethyl-1-piperazineethane sulfonic acid, n-2-hydroxyethylpiperazine-n'-ethanesulfonate
CHEBI:42334
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 230-907-9

Recommended Storage : Store at Room Temperature (15 to 30°C). Store Desiccated.

SDS
Documents

Documents

Promotions

Promotions