accessibility menu, dialog, popup
UserName
Fisher Scientific Logo
EN
chevron-down
Select Language
English
Français
Search
Order Status
Quick Order
Support
Offers
chevron-down
Featured
Spotlight
Offers
Flex Dollars
New Lab Start-up
View All
Sign In
chevron-down
chevron-down
Sign In
Don't have an account?
Create an Account
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Chemicals
Organic compounds
Unclassified Organic Compounds
MilliporeSigma™ Heparin, Sodium Salt, Porcine Intestinal Mucosa, Calbiochem™,
Structure Search
Print
MilliporeSigma™ Heparin, Sodium Salt, Porcine Intestinal Mucosa, Calbiochem™,
Catalog No.
375095100KU
Change view
Chip view icon
List view icon
Click to view available options
Quantity:
100 kU
500 kU
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
Quantity
Packaging
375095100KU
100 kU
Glass Bottle
375095500KU
500 kU
Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No.
375095100KU
Supplier
MilliporeSigma™
Supplier No.
375095100KU
Sign In
or
Register
to check your price and availability.
$146.70
/
Each
Add to Cart
Add to Cart
Description
Specifications
Product Suggestions
Videos
Documents
Certificates
Safety and Handling
Safety Services
Promotions
Specifications
Chemical Identifiers
CAS
9005-49-6
Molecular Formula
C26H42N2O37S5
Molecular Weight (g/mol)
1134.90
InChI Key
HTTJABKRGRZYRN-UHFFFAOYNA-N
Synonym
heparin, enoxaparin, clexane, lovenox, ardeparin, lmwh, lovenox hp, low molecular weight heparin, semuloparin, lipo-hepin
PubChem CID
772
Specifications
Packaging
Glass Bottle
Quantity
100 kU
Physical Form
Lyophilized
Product Suggestions
Documents