missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Heparin Sepharose™ 6 Fast Flow Affinity Chromatography Media

Catalog No. 45002970
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45002970 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45002970 Supplier Cytiva Supplier No. 17099801

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

USe for separations of biomolecules with an affinity for heparin, including antithrombin III, coagulation factors and other plasma proteins. Cytiva Life Sciences™ Heparin Sepharose™ 6 Fast Flow Affinity Chromatography Media is fast and reliable and highly suitable for process-scale purification processes.

  • Composed of cross-linked 6% agarose beads modified sodium heparin
  • Immobilization performed using chemically stable and oriented coupling giving high binding capacity
  • Enhanced binding capacity for antithrombin III due to oriented coupling of the ligand
  • Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development

Specifications

Quantity 50 mL
For Use With (Application) For Fast and Reliable Separations of Biomolecules with an Affinity for Heparin, Including Antithrombin III, Coagulation Factors and Other Plasma Proteins Highly Suitable for Process-Scale PurifiCatIon Processes
Stability Stable in commonly used aqueous Buffers: 0.1 M NaOH, 0.05 M Sodium Acetate, 8 M Urea, 6 M Guanidine Hydrochloride, non-ionic detergents, 10% Glycerol
Storage Requirements 4°C to 30°C
Product Type Heparin Sepharose 6 Fast Flow Media
Flow Rate 250 to 400 cm/Hoμr
Particle Size 45 to 165 μm
Height (English) Bed 9.84 in.
Height (Metric) Bed 25 cm
Max. Pressure 100 kPa
Matrix 6% Cross-Linked Agarose
pH Range 4 to 12
Storage Buffer 20% Ethanol, 0.05 M Sodium Acetate
Ligand Type Porcine heparin
Show More Show Less