missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ HepaRG™ Serum-free Induction Medium Supplement

Catalog No. HPRG650
Encompass
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
HPRG650 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. HPRG650 Supplier Gibco™ Supplier No. HPRG650

HepaRG™ Serum-free Induction Medium Supplement

HepaRG™ Serum-free Induction Medium Supplement is a specialized media supplement for use in culturing HepaRG™ cells. HepaRG™ Serum-free Induction Medium Supplement is specifically used for induction of metabolic activity in HepaRG™ cells in a serum free environment.
  • Easy media preparation, just thaw, mix, and use
  • Reproducible, and biologically relevant results with HepaRG™ cells

    Media Preparation
    Working media is prepared by the addition of thawed HepaRG™ Serum-free Induction Medium Supplement to 100 ml of William’s Medium E (A1217601 or 12551032) and 1 ml of GlutaMAX™-I (35050061). The simple two-step process helps reduce contamination opportunities.

    Simplified, Relevant and Reproducible Studies
    HepaRG™ cells exhibit many characteristics of primary human hepatocytes and are essentially a single donor. These features allow users to obtain physiological relevant results for metabolism-based drug-drug interactions without the concern of donor variability and limited lot sizes that come with relying on donor tissue. (Note: The cells we provide are terminally differentiated).

    HepaRG™ is a trademark of BioPredic International.
  • Specifications

    Content And Storage Supplement should be stored at -20°C
    Cell Type HepaRG Cells
    Classification Serum-free
    Form Frozen
    Product Type Hepatocyte Supplement
    Serum Level Serum-Free
    Sterility Sterile
    Without Additives No Glutamine
    For Use With (Application) Drug Metabolism
    Product Line Gibco
    Purity or Quality Grade Research Grade, Cell Culture Grade
    Quantity 2 mL
    Shipping Condition Dry Ice
    Source Serum-Free
    Tested For Sterility Testing (Bacteria & Fungi)
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.