missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ HepaRG™ Induction Medium Supplement

Catalog No. HPRG640
Encompass
Change view
Click to view available options
Quantity:
1 Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
HPRG640 1 Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. HPRG640 Supplier Gibco™ Supplier No. HPRG640

HepaRG™ Induction Medium Supplement

HepaRG™ Induction Medium Supplement is a specialized media supplement for use in culturing HepaRG™ cells. HepaRG™ Induction Medium Supplement is specifically used for induction of metabolic activity in HepaRG™ cells.
  • Easy media preparation, just thaw, mix, and use
  • Reproducible, and biologically relevant results with HepaRG™ cells

    Media Preparation
    Working media is prepared by the addition of thawed HepaRG™ Induction Medium Supplement to 100 ml of William’s Medium E (A1217601 or 12551032) and 1 ml of GlutaMAX™-I (35050061). The simple two-step process helps reduce contamination opportunities.

    Simplified, Relevant and Reproducible Studies
    HepaRG™ cells exhibit many characteristics of primary human hepatocytes and are essentially a single donor. These features allow users to obtain physiological relevant results for metabolism-based drug-drug interactions without the concern of donor variability and limited lot sizes that come with relying on donor tissue. (Note: The cells we provide are terminally differentiated).

    HepaRG™ is a trademark of BioPredic International.
  • Specifications

    Content And Storage Supplement should be stored at -20°C
    Cell Type HepaRG Cells
    Form Frozen
    Product Type Hepatocyte Supplement
    Serum Level Standard Serum
    Sterility Sterile
    Without Additives No Glutamine
    For Use With (Application) Drug Metabolism
    Product Line Gibco
    Purity or Quality Grade Research Grade, Cell Culture Grade
    Quantity 1 Bottle
    Shipping Condition Dry Ice
    Tested For Sterility Testing (Bacteria & Fungi)
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.