missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HEPACAM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116318
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116318 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116318 Supplier Invitrogen™ Supplier No. PA5116318

Rabbit Polyclonal Antibody

HEPACAM2 Polyclonal Antibody for Western Blot

HEPACAM2 (Hepatocyte cell adhesion molecule 2), a type I N-linked transmembrane glycoprotein, belongs to the immunoglobulin superfamily. The exact function of HEPACAM2 is currently unknown, but the related protein HEPACAM forms cishomodimers on the cell surface to regulate the cell adhesion and may inhibit cell growth through suppression of cell proliferation. HEPACAM and HEPACAM2 mRNA are differentially regulated in canine tumors, with HEPACAM2 mRNA showing increased levels in adenomas, but decreased levels in metastatic carcinomas compared to normal tissues, while HEPACAM protein levels decreased in adenomas. It is therefore likely that HEPACAM2 plays a different role than HEPACAM in the development and progression to tumors.

Specifications

Antigen HEPACAM2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene HEPACAM2
Gene Accession No. A8MVW5
Gene Alias AI987662; HEPACAM family member 2; HEPACAM2; Miki; Mitotic kinetics regulator; RGD1305697; UNQ305/PRO346
Gene Symbols HEPACAM2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 371-400 amino acids from the C-terminal region of human HEPACAM2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 253012
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less