missing translation for 'onlineSavingsMsg'
Learn More

Hentriacontane, ≥98% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014970
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014970 250 mg
111014969 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014970 Supplier Merck Emd Millipore Supplier No. 51529250MG
Only null left

Hentriacontane is a normal alkane, comprising of approximately 31 carbon atoms, which can be found in beewax. It is also found to be a major component of candelilla wax, which upon organo-gelation can provide structure to solid-like fat-based foods, drug delivery systems and cosmetics.

Featured Industries: Petroleum

Specifications

CAS 630-04-6
Physical Form Neat
Linear Formula CH3(CH2)29 CH3
MDL Number MFCD00048745
Melting Point 67°C to 69°C
Type Analytical Standard
Quantity 250 mg
Formula Weight 436.84
Percent Purity ≥98% (GC)
Grade Analytical Standard