missing translation for 'onlineSavingsMsg'
Learn More

Heneicosane, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014957
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014957 1 g
111014958 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014957 Supplier Merck Emd Millipore Supplier No. 515231G
Featured Industries: Petroleum

Specifications

CAS 629-94-7
Vapor Pressure <1 mmHg (20°C)
Vapor Density 10.3 (vs. air)
Flash Point 113°C
Physical Form Neat
Linear Formula CH3(CH2)19 CH3
MDL Number MFCD00009346
Melting Point 39°C to 41°C; 39°C to 42°C
Boiling Point 100°C (2 mmHg); 356.1°C (760 mmHg)
Type Analytical Standard
Quantity 1 g
Formula Weight 296.57
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less