missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Hemoglobin Monoclonal Antibody (HB11-201.11), Invitrogen™

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA110806
Description
MA1-10806 detects Hemoglobin from human samples. This antibody does not detect bovine, chicken, equine, ovine, or porcine Hemoglobin. MA1-10806 has been successfully used in ELISA and Western blot applications. The MA1-10806 immunogen is purified human hemoglobin.
The human alpha globin gene cluster located on chromosome 16 spans about 30 kb and includes seven loci: 5'- zeta - pseudozeta - mu - pseudoalpha-1 - alpha-2 - alpha-1 - theta - 3'. The alpha-2 (HBA2) and alpha-1 (HBA1) coding sequences are identical. These genes differ slightly over the 5' untranslated regions and the introns, but they differ significantly over the 3' untranslated regions. Two alpha chains plus two beta chains constitute HbA, which in normal adult life comprises about 97% of the total hemoglobin; alpha chains combine with delta chains to constitute HbA-2, which with HbF (fetal hemoglobin) makes up the remaining 3% of adult hemoglobin. Alpha thalassemias result from deletions of each of the alpha genes as well as deletions of both HBA2 and HBA1; some nondeletion alpha thalassemias have also been reported.Chemical Identifiers
| Hemoglobin | |
| 1.4 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.4 | |
| alpha one globin; alpha-1 globin; alpha-1-globin; alpha-2 globin chain; Alpha-globin; ALPHA-GLOBIN GENE; delta globin; Haemoglobin; Hb; HBA1; HbA1c; HBA2; HBA-T3; HBH; hemoglobin alpha 1 globin chain; Hemoglobin alpha chain; Hemoglobin subunit alpha; hemoglobin subunit alpha 1; hemoglobin, alpha 1 | |
| HBA1, HBB | |
| IgG1 | |
| Protein G | |
| Antibody |
| HB11-201.11 | |
| ELISA, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P68871, P69905 | |
| 3039, 3043 | |
| Purified human hemoglobin. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| HBA1 |
Specifications
| Hemoglobin | |
| Monoclonal | |
| 1.4 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| P68871, P69905 | |
| HBA1, HBB | |
| Purified human hemoglobin. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot, Immunocytochemistry | |
| HB11-201.11 | |
| Unconjugated | |
| HBA1 | |
| alpha one globin; alpha-1 globin; alpha-1-globin; alpha-2 globin chain; Alpha-globin; ALPHA-GLOBIN GENE; delta globin; Haemoglobin; Hb; HBA1; HbA1c; HBA2; HBA-T3; HBH; hemoglobin alpha 1 globin chain; Hemoglobin alpha chain; Hemoglobin subunit alpha; hemoglobin subunit alpha 1; hemoglobin, alpha 1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 3039, 3043 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |