Learn More
VELP Scientifica™ Hemispheric Bowls for Round Bottom Flasks
The Hemispheric Bowl is a suitable solution for replacing oil baths and mantles in order to perform clean and safe analysis with round bottom flasks.
$574.56 - $3620.99
Specifications
| For Use With (Equipment) | Round Bottom Flasks |
|---|---|
| Type | Flask Bowl |
| Coating | Aluminum |
| Material | Aluminum |
Description
- Precisely fits 135 mm diameter plate
- Accommodates different sizes of round bottom flasks
- Minimizes risk of movement
- Ensures fastest heat transfer
Specifications
| Round Bottom Flasks | |
| Flask Bowl | |
| Aluminum | |
| Aluminum |