Learn More
Hematoxylin Stain Solution, Gill 2 Formulation, Double Strength, Ricca Chemical
For More Intense Nuclei in Progressive Cytology and Delicate Nuclei in Progressive Histology
Supplier: Les Produits Chimiques 8255725
Specifications
| Hematoxylin Stain Solution | |
| 7732-18-5,107-21-1,64-19-7,7784-31-8,517-28-2,7681-55-2 | |
| 75.0,20.88,3.15,2.64,0.3,0.03 | |
| Cubitainer | |
| 2 to 4 | |
| Liquid |
| Gill 2 Formulation, Double Strength | |
| 72.0,20.06,3.03,2.54,0.29,0.03 | |
| Laboratory | |
| Dark Purple | |
| 20 L |