missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hematoxylin Hydrate 97.0+%, TCI America™
$134.80 - $398.70
Chemical Identifiers
| CAS | 517-28-2 |
|---|---|
| Molecular Formula | C16H14O6 |
| Molecular Weight (g/mol) | 302.28 |
| MDL Number | MFCD00078111 |
| InChI Key | WZUVPPKBWHMQCE-WKTCHCBJNA-N |
| Synonym | Natural Black 1, Haematoxylin |
| PubChem CID | 45029742 |
| IUPAC Name | (1R,10S)-8-oxatetracyclo[8.7.0.0²,⁷.0¹²,¹⁷]heptadeca-2(7),3,5,12(17),13,15-hexaene-5,6,10,14,15-pentol |
| SMILES | [H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3 |
Chemical Identifiers
| 517-28-2 | |
| 302.28 | |
| WZUVPPKBWHMQCE-WKTCHCBJNA-N | |
| 45029742 | |
| [H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3 |
| C16H14O6 | |
| MFCD00078111 | |
| Natural Black 1, Haematoxylin | |
| (1R,10S)-8-oxatetracyclo[8.7.0.0²,⁷.0¹²,¹⁷]heptadeca-2(7),3,5,12(17),13,15-hexaene-5,6,10,14,15-pentol |
Specifications
| 517-28-2 | |
| MFCD00078111 | |
| WZUVPPKBWHMQCE-WKTCHCBJNA-N | |
| (1R,10S)-8-oxatetracyclo[8.7.0.0²,⁷.0¹²,¹⁷]heptadeca-2(7),3,5,12(17),13,15-hexaene-5,6,10,14,15-pentol | |
| 45029742 | |
| ≥97.0% (GC) | |
| Hematoxylin Hydrate |
| C16H14O6 | |
| Natural Black 1, Haematoxylin | |
| [H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3 | |
| 302.28 | |
| 302.28 | |
| Crystal/Powder |
Safety and Handling
EINECSNumber : (5)-3664
RTECSNumber : MH7875000
TSCA : Yes