Learn More
Heidolph™ Overhead Stirrer Impellers - Propeller Style
Hei-TORQUE Propeller Style Impellers are recommended for applications which require average or high speed and medium to high viscosity; excellent mixing for homogenization and suspensions
Supplier:  Heidolph™ 036300410
 View more versions of this product
Catalog No. 13880135
Only null left
Description
Description
  • Excellent mixing properties for homogenization and suspensions
  • For mixing tasks with medium or high viscosity
  • Axial flow
  • Made from PTFE or stainless-steel
Specifications
Chemical Identifiers
0.31 in.
Mixing tasks with medium or high viscosity, excellent mixing properties for homogenization and suspensions
48 oz.
0.8 cm
Propeller Impeller
3 lb.
Specifications
1.3 in.
3.3 cm
Mixing tasks with medium or high viscosity, excellent mixing properties for homogenization and suspensions
400 mm
48 oz.
0.31 in.
0.8 cm
15.74 in.
Propeller Impeller
3 lb.
Product Suggestions
Product Suggestions