Learn More
Heidolph™ Radleys™ Reaction Vessel Accessory: DN200 Lids
Catalog No. 13890016
Change view
Click to view available options
Type:
6-Neck Lid
7-Neck Lid
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
13890016 7-Neck Lid
13890015 6-Neck Lid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13890016 Supplier Heidolph™ Supplier No. 015908006
Glass lids with Rodaviss conical ground glass joints
  • For reaction vessels used in Reactor-Ready™ Pilot Lab Reactor
  • Lid with grease free Rodaviss joints that are interchangeable with ordinary 1-in-10 B and A length tapered conical joints and will not jam
  • Available as 6 or 7 neck
Inlets in 6 Neck DN200 Lid
  • 1 x B34 central vertical - stirrer
  • 1 x B19 side vertical - probe
  • 1 x B24 side vertical - gas purge
  • 1 x B29 side vertical - dropping funnel, bubbler
  • 1 x B34 side vertical - condenser and reflux
  • 1 x B45 side angled - solid additions
Inlets in 7 Neck DN200 Lid
  • 1 x B34 central vertical - stirrer
  • 1 x B19 side vertical - probe
  • 1 x B24 side vertical - gas purge
  • 1 x B29 side vertical - drop funnel, bubbler
  • 2 x B34 side vertical - condenser and reflux
  • 1 x B45 side angled - solid additions
Specifications
Type 7-Neck Lid
For Use With (Equipment) Reaction Vessel