Learn More
Heidolph™ Radleys Carousel 6 Plus Reaction Station™ Packages
Designed to be highly flexible, stations are configured with a combination of flask sizes with different sidearm combinations and accessories
Supplier: Heidolph™ 015850460
Description
The Carousel itself fits on a standard hotplate stirrer and is compatible with standard rotary and most centrifugal evaporators.Various combinations available for both heated and cooled reactions. See Specifications tab for package details
Heated Unit
With an operating temperature range from ambient to 180°C, a water-cooled aluminum reflux head can also provide efficient refluxing within individual glass reflux necks; combine a radial gas distribution system with gas-tight PTFE caps, and you can perform air-sensitive reactions under inert atmospheric conditions.
- Solid aluminum base for rapid and even heating of all flasks
- Fuzzy logic temperature controller is accurate to ±0.5°C
- Uses the single-rotating magnetic field of the hotplate stirrer to stir all 6 round-bottom flasks evenly and powerfully
- PTFE guard-ring protects against accidental contact with hot base
- Removable aluminum inserts in the flux head allow easy removal of flasks and maintain good heat transfer for cooling (built-in spring clip keeps flasks in position once inserts are removed)
- Acetal quick-release valve couplings allow you to disconnect the reflux head without losing any of the cooling water
With an operating temperature range from ambient down to -78°C, you can use any combination of flask sizes in the cooled Carousel.
- Carousel design and technology provides even cooling and efficient stirring for samples
- HDPE cooling reservoir is compatible with a wide range of freezing mixtures, including dry ice/acetone
- Insulated foam core maintains low temperatures for long periods, reducing condensation and ice formation on outer surfaces while it protects the stirrer from freezing
- Stainless-steel central inlet/outlet for vacuum and inert gas, radial gas distribution system and gastight PTFE caps for reactions under inert atmosphere
- Carousel protective cover reduces frost formation, maintains temperature of cooling mixture, and reduces risk of solvent spitting
Specifications
| Carousel 6 plus Reaction Station, easy on PTFE cap, carousel tech stirring hotplate, Pt 1000, Silicone septa for PTFE caps, PTFE Magnetic stirring bar retriever 350 mm, 250 mL reaction flask, reflux tube, connection, B24/29 rodaviss sealing cap, rotary evaporator adapters ordinary B29 socket to B24 cone, reaction flask support ring, elliptical stirring bar 25 mm RE, tubing for inert gas or reflux cooling, 250 mL reaction flasks with B24/29 sidearm and septa port, 2 x B24/29 liquid additions dropping funnel and pressure equalising arm, B24/29 solid additions funnel, PTFE/silicone septa 29 mm | |
| System 35; Heated | |
| Carousel 6 Plus Reaction Station Package |