missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heidolph™ Laborota™ 20 Distimatic Rotary Evaporators
For unattended and continuous operation including automatic release of residue, automatic module is compatible with all Labrota 20 models
$109,464.87
Specifications
| For Use With (Application) | Unattended and continuous operation including automatic release of residue |
|---|---|
| Type | Distimatic Rotary Evaporator |
| Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
|
13889371
|
Heidolph™
036064600 |
Adapter piece of Glassware set A, sensor |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Save time by distilling large volumes in a continuous and unattended way overnight and during the weekend:
- Automatic refill of evaporation flask with solvent mixture set according to your settings and continuous automatic release of condensate
- Unique automatic residue drainage offers wide range of new possibilities for automatic distillation
- All parameters can be set individually
- Automatic module is compatible with all Laborota 20 models and also with other comparable bench top evaporators
- Storage and the collecting tank can be replaced during the process
- Valve matrix controls continuous drainage of condensate from the system into a temporary storage flask; afterwards the collected condensate will be released into collecting tank by a diaphragm pump
- Filling level in evaporating flask is measured by a sensor
- Required filling level can be set individually
- Recharge of the product under vacuum is valve-controlled
- Sensor measures the filling level of the storage tank and stops the distillation process as soon as the liquid is completely evaporated
Automatic release of residue:
- Residue in evaporating flask will be released into a collecting tank; afterwards the process starts again automatically by refilling the evaporating flask
- Process can be terminated automatically after evaporation is completed or manually at any time by the operator
Specifications
| Unattended and continuous operation including automatic release of residue | |
| Distimatic Rotary Evaporator |