missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ Laborota™ 20 Distimatic Rotary Evaporators

Catalog No. 13889371
Change view
Click to view available options
Includes:
Adapter piece of Glassware set A, sensor
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
13889371 Adapter piece of Glassware set A, sensor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13889371 Supplier Heidolph™ Supplier No. 036064600

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For unattended and continuous operation including automatic release of residue, automatic module is compatible with all Labrota 20 models

Save time by distilling large volumes in a continuous and unattended way overnight and during the weekend:

  • Automatic refill of evaporation flask with solvent mixture set according to your settings and continuous automatic release of condensate
  • Unique automatic residue drainage offers wide range of new possibilities for automatic distillation
  • All parameters can be set individually
  • Automatic module is compatible with all Laborota 20 models and also with other comparable bench top evaporators
  • Storage and the collecting tank can be replaced during the process
  • Valve matrix controls continuous drainage of condensate from the system into a temporary storage flask; afterwards the collected condensate will be released into collecting tank by a diaphragm pump
Automatic Solvent supply of the evaporating flask:
  • Filling level in evaporating flask is measured by a sensor
  • Required filling level can be set individually
  • Recharge of the product under vacuum is valve-controlled
  • Sensor measures the filling level of the storage tank and stops the distillation process as soon as the liquid is completely evaporated

Automatic release of residue:

  • Residue in evaporating flask will be released into a collecting tank; afterwards the process starts again automatically by refilling the evaporating flask
End of distillation process:
  • Process can be terminated automatically after evaporation is completed or manually at any time by the operator

Specifications

For Use With (Application) Unattended and continuous operation including automatic release of residue
Includes Adapter piece of Glassware set A, sensor
Type Distimatic Rotary Evaporator