Learn More
Heidolph™ Carousel Attachments for Multi Reax Test Tube Shaker
Accessory for Multi Reax Test Tube Shakers
Supplier: Heidolph™ 036303530
Description
Carousel test tube attachment for 26 vessels with diameters from 10 to 16 mm part number 036303530 included with purchase of Multi Reax.
Specifications
| Test Tubes | |
| Test Tube Carousel Attachment | |
| Multi Reax Test Tube Shaker |
| 10 to 16 mm | |
| 160 mm | |
| 12 |