Learn More
Sol-Care™ Heel Blood Lancets
Heel Blood Lancet is sterile, single use medical device use to collect blood samples from the heels.
Supplier: Sol-Care™ LAH004
Description
- Press trigger allows for one hand activation
- Visual indicator assists in puncture precision
- Unexposed blade prevents accidental needle stick injuries
- Safety stop prevents accidental misfiring
Chemical Identifiers
| Heel Incision Device | |
| Single-use |
Specifications
| Heel Incision Device | |
| Single-use | |
| 1.5 x 3 mm |