Learn More
Sol-Care™ Heel Blood Lancets
Catalog No. p-200148529
Change view
Click to view available options
Size:
0.65 x 1.5 mm
0.85 x 1.75 mm
1.5 x 3 mm
1 x 2.5 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
361060371 0.65 x 1.5 mm
361060361 0.85 x 1.75 mm
361060244 1 x 2.5 mm
361060360 1.5 x 3 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 361060371 Supplier Sol-Care™ Supplier No. LAH001
Only null left
Heel Blood Lancet is sterile, single use medical device use to collect blood samples from the heels.
  • Press trigger allows for one hand activation
  • Visual indicator assists in puncture precision
  • Unexposed blade prevents accidental needle stick injuries
  • Safety stop prevents accidental misfiring
Specifications
Product Type Heel Incision Device
Disposable Single-use
Size 0.65 x 1.5 mm