missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hederacoside C, MilliporeSigma™ Supelco™
Description
Food and Beverages
Specifications
Specifications
| Packaging | Bottomless Glass Bottle with Inserted Fused Cone |
| CAS | 14216-03-6 |
| Physical Form | Neat |
| Linear Formula | C59H96O26 |
| MDL Number | MFCD00198062 |
| Melting Point | 222°C |
| Type | Analytical Standard |
| Quantity | 50 mg |
| Formula Weight | 1221.38 |
| Grade | Analytical Standard |