missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HECTD2 Monoclonal Antibody (K24026_13A9), Invitrogen™

Catalog No. MA555809
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA555809 50 μL
MA556189 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA555809 Supplier Invitrogen™ Supplier No. MA555809
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: PEERKGKESE REKLPPIVSA GAGATAGLDR GAKGQISTFS SFISAVSPKK EAAENRSSPA HLVFPNIKNV REPPPICLDV RQKQRTSMDA SSSEMKAPVL PEPILPIQPK TVKDFQEDVE KVKSSGDWKA VHDFYLTTFD SFPELNAAFK

Probable E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates.

Specifications

Antigen HECTD2
Applications Western Blot
Classification Monoclonal
Clone K24026_13A9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene Hectd2
Gene Accession No. Q5U5R9
Gene Alias 4921524L07; A630025O09Rik; AW212605; HECT domain containing 2; HECT domain containing E3 ubiquitin protein ligase 2; HECT domain E3 ubiquitin protein ligase 2; HECT domain-containing protein 2; Hectd2; HECT-type E3 ubiquitin transferase HECTD2; hmm77879; Probable E3 ubiquitin-protein ligase HECTD2
Gene Symbols Hectd2
Host Species Mouse
Immunogen Recombinant human HECTD2 (immunogen range is 21-170).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143279
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less