missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heavy Metals, plasma standard solution, Specpure™, Ag, As, Ba, Cd, Cr, Hg, Pb, Se @ 100μg/mL
$326.58 - $326.58
Specifications
| Chemical Name or Material | Heavy Metals, plasma standard solution |
|---|---|
| Name Note | Ag, As, Ba, Cd, Cr, Hg, Pb, Se @ 100μg/mL |
| Physical Form | Liquid |
| Concentration | Matrix: 5% HNO3 |
| MDL Number | MFCD02091713 |
Description
Heavy Metals, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Heavy Metals, plasma standard solution | |
| Liquid | |
| MFCD02091713 | |
| Miscible with water |
| Ag, As, Ba, Cd, Cr, Hg, Pb, Se @ 100μg/mL | |
| Matrix: 5% HNO3 | |
| UN3264 | |
| Specpure |
RUO – Research Use Only