missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heavy Magnesium Oxide, Heavy, Powder, FCC, 96-100.5%, Spectrum™ Chemical
M1168, 1309-48-4, MgO
$653.10 - $8416.63
Chemical Identifiers
| CAS | 1309-48-4 |
|---|---|
| Molecular Formula | MgO |
| Molecular Weight (g/mol) | 40.30 |
| MDL Number | MFCD00011109 |
| InChI Key | CPLXHLVBOLITMK-UHFFFAOYSA-N |
| IUPAC Name | oxomagnesium |
| SMILES | O=[Mg] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18603097
|
Spectrum Chemical Mfg Cor
M1168500GM |
500 g | Poly Bottle |
Each for $653.10
|
|
||||
|
18603098
|
Spectrum Chemical Mfg Cor
M116825KG |
2.5 kg | Poly Pail |
Each for $1,851.93
|
|
||||
|
18603099
|
Spectrum Chemical Mfg Cor
M116812KG |
12 kg | Poly Pail |
Each for $4,026.50
|
|
||||
|
18603100
|
Spectrum Chemical Mfg Cor
M116825KGBL |
25 kg | Fiber Drum |
Each for $8,416.63
|
|
||||