missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Heating Blocks for Microtube Shaker
These nine interchangeable aluminum blocks accommodate PCR plates and a variety of common life science vessels.
Supplier: Axygen™ I4010E
Description
- No additional tools required for block exchanges
- Blocks utilize magnetic adhesion technology and are self-recognized once exchanged
- Combination of heating, cooling, and mixing with versatile block selection makes the Axygen™ Microtube Shaker ideal for many life science laboratories
- Applications include but are not limited to gene synthesis, gene purification and denaturation, enzymatic reactions, and bacterial growth assays
Chemical Identifiers
| Aluminum | |
| Axygen™ Microtube Shaker |
Specifications
| 1400 rpm | |
| Aluminum | |
| Cone Bottom |
| 5 mL | |
| Axygen™ Microtube Shaker | |
| 8-well |