Learn More
Axygen™ Heating Blocks for Microtube Shaker
These nine interchangeable aluminum blocks accommodate PCR plates and a variety of common life science vessels.
Supplier:  Axygen™ I4010B
 View more versions of this product
Catalog No. 14223087
Only null left
Description
Description
  • No additional tools required for block exchanges
  • Blocks utilize magnetic adhesion technology and are self-recognized once exchanged
  • Combination of heating, cooling, and mixing with versatile block selection makes the Axygen™ Microtube Shaker ideal for many life science laboratories
  • Applications include but are not limited to gene synthesis, gene purification and denaturation, enzymatic reactions, and bacterial growth assays
Specifications
Chemical Identifiers
Aluminum
Axygen™ Microtube Shaker
Specifications
1500 rpm
Aluminum
Cone Bottom
1.5 mL
Axygen™ Microtube Shaker
24-well
Product Suggestions
Product Suggestions