missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BUCHI Heating Bath B-305
Compatible with evaporating flasks up to 5 liters.
Supplier: BUCHI 11B305102
Description
- Use with water or oil as heating media
- For temperatures up to 220°C
Specifications
| Stainless Steel | |
| Water or Oil | |
| Heating Bath (B-305), Base (B-300) | |
| 5.5 L |
| Heating Bath | |
| R-300 | |
| ≤220°C |