missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mopec™ Heated Forceps System
The Heated Forceps System is designed to take up minimal bench top space and comes complete with a power supply, control unit, and two different heated forceps.
$6989.80 - $6989.80
| Catalog Number | Mfr. No. | Tip Style | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Tip Style | Price | Quantity | |||||
|
22-445-423
|
Mopec™
BK300SE |
Serrated |
Each for $6,989.80
|
|
|||||
|
22-445-524
|
Mopec™
BK300SM |
Smooth |
Each for $6,989.80
|
|
|||||
Description
- Ergonomic design ensures user comfort while performing specimen placement, and the anti-microbial coating helps prevent bacteria and virus growth
- Choose either serrated or smooth tips
- System includes: 1 mm Heated Forceps (Red), 2 mm Heated Forceps (Yellow), Mopec™ Control Unit, Mopec™ Power Supply
- Designed to take up minimal bench-top space
- Anti-microbial coating to help prevent bacteria and virus growth
- Forceps operate at a temperature between 63°C to 70°C (145°F to 158°F)
Specifications
| 145°F to 158°F |