missing translation for 'onlineSavingsMsg'
Learn More

HEAB Recombinant Rabbit Monoclonal Antibody (23GB3085)
GREENER_CHOICE

Catalog No. MA553596
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553596 100 μL
MA553597 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553596 Supplier Fisher Chemical Supplier No. MA553596

HEAB Recombinant Rabbit Monoclonal Antibody (23GB3085)

HEAB Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Specifications

Antigen HEAB
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3085
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CLP1
Gene Accession No. Q92989
Gene Alias AI462438; ATP/GTP-binding protein; cleavage and polyadenylation factor I subunit 1; CLP1; CLP1, cleavage and polyadenylation factor I subunit; CLP1, cleavage and polyadenylation factor I subunit, homolog; HAMAP-Rule:MF_03035}; hClp1; Heab; homolog of yeast CFIA subunit Clp1p; polyadenylation factor Clp1; polyadenylation factor Clp1 {ECO:0000255; Polynucleotide kinase Clp1; polynucleotide kinase Clp1 {ECO:0000255; polyribonucleotide 5'-hydroxyl-kinase Clp1; polyribonucleotide 5'-hydroxyl-kinase Clp1 {ECO:0000255; Pre-mRNA cleavage complex II protein Clp1; pre-mRNA cleavage complex II protein Clp1 {ECO:0000255; RGD1307679
Gene Symbols CLP1
Host Species Rabbit
Immunogen A synthesized peptide derived from HEAB (1-25AA).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10978
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less