missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HDLBP Recombinant Rabbit Monoclonal Antibody (23GB3970)

Catalog No. MA552649
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552649 20 μL
MA552648 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552649 Supplier Invitrogen™ Supplier No. MA552649
Only null left

Rabbit Recombinant Monoclonal Antibody

HDLBP Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

High density lipoprotein-binding protein, also known as vigilin, is a 110-kD protein that specifically binds HDL molecules and may function in the removal of excess cellular cholesterol.

Specifications

Antigen HDLBP
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3970
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HDLBP
Gene Accession No. Q00341, Q8VDJ3, Q9Z1A6
Gene Alias 1110005P14Rik; AA960365; AI118566; D1Ertd101e; HBP; HDL binding protein; HDL-binding protein; HDLBP; Hemopexin; high density lipoprotein (HDL) binding protein; high density lipoprotein binding protein; high density lipoprotein binding protein (vigilin); high density lipoprotein-binding protein; Hpx; Hpxn; hx; lipoprotein-binding protein; PRO2900; VGL; vigilin
Gene Symbols HDLBP
Host Species Rabbit
Immunogen A synthesized peptide derived from human HDLBP (70-100AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110611, 3069, 64474
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less