missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HDGFRP2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA521171

Catalog No. PIPA521171


Only null left

Description

Description

A suggested positive control is human testis tissue lysate. PA5-21171 can be used with blocking peptide PEP-1285.

Hepatoma-derived growth factor-related protein 2 (HDGFRP2) is a widely expressed member of protein family that contains the HDGF related proteins 1-4 and lens epithelial derived growth factor (LEDGF). These family members show extensive homology at the amino terminus (termed the hath region). Little is known of the role of HDGFRP2, but like the related LEDGF, HDGFRP2 can bind HIV-1 integrase and stimulate HIV-1 integrase activity in vitro.
Specifications

Chemical Identifiers

HDGFRP2
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Polyclonal
Liquid
IgG
Human, Mouse, Rat
HDGFL2
HDGF2; HDGF-2; Hdgf3; HDGF-3; Hdgfl2; HDGF-related protein 2; HDGFRP2; Hepatoma-derived growth factor 2; Hepatoma-derived growth factor 3; hepatoma-derived growth factor, related protein 2; hepatoma-derived growth factor-related protein 2; HRP2; HRP-2; UNQ785/PRO1604
HDGFL2
Primary
Antigen affinity chromatography
1 mg/mL
Immunohistochemistry, Western Blot
Unconjugated
Rabbit
RUO
PBS with 0.02% sodium azide
Q3UMU9, Q7Z4V5, Q925G1
15193, 171073, 84717
An 18 amino acid peptide near the amino terminus of human HDGFRP2.
Antibody

Specifications

HDGFRP2
Polyclonal
Unconjugated
HDGFL2
HDGF2; HDGF-2; Hdgf3; HDGF-3; Hdgfl2; HDGF-related protein 2; HDGFRP2; Hepatoma-derived growth factor 2; Hepatoma-derived growth factor 3; hepatoma-derived growth factor, related protein 2; hepatoma-derived growth factor-related protein 2; HRP2; HRP-2; UNQ785/PRO1604
Rabbit
Antigen affinity chromatography
RUO
15193, 171073, 84717
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Liquid
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q3UMU9, Q7Z4V5, Q925G1
HDGFL2
An 18 amino acid peptide near the amino terminus of human HDGFRP2.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions