missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HDGF Recombinant Mouse Monoclonal Antibody (H3)

Description
HDGF Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC, Flow, IP, Neu
Specifications
Specifications
| Antigen | HDGF |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Neutralization, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | H3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% ProClin 300 |
| Gene | HDGF |
| Gene Accession No. | P51858, P51859 |
| Gene Alias | AI118077; D3Ertd299e; DKFZp686J1764; FLJ96580; Hdgf; Hepatoma-derived growth factor; high mobility group protein 1-like 2; HMG1L2; HMG-1L2; Tdrm1 |
| Show More |