missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HDAC11 Recombinant Rabbit Monoclonal Antibody (23GB6005)

Catalog No. MA552971
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552971 20 μL
MA552970 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552971 Supplier Invitrogen™ Supplier No. MA552971
Only null left

Rabbit Recombinant Monoclonal Antibody

HDAC11 Recombinant Monoclonal Antibody for ICC/IF, Western Blot

This gene encodes a class IV histone deacetylase. The encoded protein is localized to the nucleus and may be involved in regulating the expression of interleukin 10. Alternative splicing results in multiple transcript variants.

Specifications

Antigen HDAC11
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB6005
Concentration 0.56 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HDAC11
Gene Accession No. Q91WA3, Q96DB2
Gene Alias FLJ22237; HD11; HDAC11; Histone deacetylase 11
Gene Symbols HDAC11
Host Species Rabbit
Immunogen A synthesized peptide derived from human HDAC11 (205-269AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 232232, 79885
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less