missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HCS CellMask™ Stains

Catalog No. H32722
Encompass
Change view
Click to view available options
Description:
HCS CellMask Blue Stain
HCS CellMask Deep Red Stain
HCS CellMask Green Stain
HCS CellMask Near-IR Stain
HCS CellMask Orange Stain
HCS CellMask™ Red Stain
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
H32722 HCS CellMask Near-IR Stain
H32720 HCS CellMask Blue Stain
H32714 HCS CellMask Green Stain
H32713 HCS CellMask Orange Stain
H32712 HCS CellMask™ Red Stain
H32721 HCS CellMask Deep Red Stain
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. H32722 Supplier Invitrogen™ Supplier No. H32722

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Easily visualize entire cells or individual cell structures during high-content screening (HCS) assays with HCS CellMask stains, which are available in a variety of colors for multiplexing flexibility and can be used immediately after fixation and permeabilization, or after antibody labeling.

Identify entire cells (cytoplasm and nucleus) during high-content screening (HCS) assays with the versatile HCS CellMask Red, CellMask Orange, CellMask Green, CellMask Blue, CellMask Deep Red, and CellMask Near-IR stains, which are fluorophores that can be used immediately after fixation and permeabilization, or after antibody labeling. They are available in a variety of colors for multiplexing protocols.

The versatile HCS CellMask Red, CellMask Orange, CellMask Green, CellMask Blue, CellMask Deep Red, and CellMask Near-IR stains are cell delineation tools for high content screening (HCS) platforms, label the entire cell (cytoplasm and nucleus) and provide an accurate backdrop against which the features of interest can be assessed. The HCS CellMask Red and Blue stains replace the HCS CellMask Red cytoplasmic/nuclear (Cat. No. H32711) and HCS CellMask Blue cytoplasmic/nuclear stains (Cat. No. H34558), respectively.

HCS CellMask stains can be applied to cells immediately after fixation or in the last step of multiplexing protocols, and they are compatible with detergent-based cell permeabilization protocols. If only nuclear staining is desired, HCS NuclearMask stains can be used for measuring DNA content in addition to enabling robust cell demarcation. HCS NuclearMask stains are also available in a choice of colors for multiplexing flexibility.

Specifications

Content And Storage Includes 1 set of vials, providing sufficient material to stain a total of ten 96-well microplates.

Store at ≤-20°C, desiccate, and protect from light.

Format Vial(s)
For Use With (Equipment) High Content Instrument
Quantity 1 Set
Detection Method Fluorescence
Description HCS CellMask Near-IR Stain
Product Line CellMask
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Stain
Sub Cellular Localization Cytoplasm, Cytosol, Nucleus
Show More Show Less
What is the shelf life of the HCS CellMask Near-IR Stain?

As shipped, the product has a warranty period of 1 year from the date of shipment. Once the solid stain is resolubilized in DMSO, it should be stored frozen, desiccated, and protected from light and is useable for ~ 6 months.

What are the components of the HCS CellMask Near-IR Stain and how should I store them?

Each HCS CellMask Stain provides sufficient material for labeling approximately 10 x 96-well plates using an assay volume of 100 µL. The product is shipped as two components: a single vial of 250 µg of solid Near-IR Stain (Component A) and a single vial of 100 µL of DMSO (Component B).

We recommend storing Component A at ≤ -20 degrees C, desiccated and protected from light and Component B desiccated at ≤ 25 degrees C.

What dyes are used to make the CellMask stains?

The proprietary fluorescent dyes in the CellMask stains are general cytoplasmic stains. They are not found to bind to any specific cellular component.