missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HCS CellMask™ Stains

Easily visualize entire cells or individual cell structures during high-content screening (HCS) assays with HCS CellMask stains, which are available in a variety of colors for multiplexing flexibility and can be used immediately after fixation and permeabilization, or after antibody labeling.

Supplier:  Invitrogen™ H32712

Encompass

 View more versions of this product

Catalog No. H32712


Only null left

Description

Description

Identify entire cells (cytoplasm and nucleus) during high-content screening (HCS) assays with the versatile HCS CellMask Red, CellMask Orange, CellMask Green, CellMask Blue, CellMask Deep Red, and CellMask Near-IR stains, which are fluorophores that can be used immediately after fixation and permeabilization, or after antibody labeling. They are available in a variety of colors for multiplexing protocols.

The versatile HCS CellMask Red, CellMask Orange, CellMask Green, CellMask Blue, CellMask Deep Red, and CellMask Near-IR stains are cell delineation tools for high content screening (HCS) platforms, label the entire cell (cytoplasm and nucleus) and provide an accurate backdrop against which the features of interest can be assessed. The HCS CellMask Red and Blue stains replace the HCS CellMask Red cytoplasmic/nuclear (Cat. No. H32711) and HCS CellMask Blue cytoplasmic/nuclear stains (Cat. No. H34558), respectively.

HCS CellMask stains can be applied to cells immediately after fixation or in the last step of multiplexing protocols, and they are compatible with detergent-based cell permeabilization protocols. If only nuclear staining is desired, HCS NuclearMask stains can be used for measuring DNA content in addition to enabling robust cell demarcation. HCS NuclearMask stains are also available in a choice of colors for multiplexing flexibility.

Order Info

Shipping Condition: Room temperature

Specifications

Chemical Identifiers

High Content Instrument
Fluorescence
Room Temperature
Stain
1 Set
CellMask
Fluorescent Dye
Cytoplasm, Cytosol, Nucleus

Specifications

Red
588/612
1 Set
HCS CellMask™ Red Stain
CellMask
Fluorescent Dye
Cytoplasm, Cytosol, Nucleus
Includes 1 set of vials, providing sufficient material to stain a total of ten 96-well microplates.

Store at ≤-20°C, desiccate, and protect from light.

High Content Instrument
Fluorescence
Visible
Room Temperature
Stain
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions