missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HCLS1 Recombinant Rabbit Monoclonal Antibody (PSH01-12)

Description
Sequence Similarities: 70% Mouse. Tissue Specificity: Expressed only in tissues and cells of hematopoietic origin. Expressed in early stage of myeloid and erythroid differentiation. Positive Control: Ramos cell lysate, Daudi cell lysate, Raji cell lysate, Jurkat cell lysate, HL-60 cell lysate, human lung carcinoma tissue, human liver tissue, HL-60, Jurkat. Subcellular Location: Membrane, Cytoplasm, Mitochondrion. Predicted band size: 54 kDa.
Substrate of the antigen receptor-coupled tyrosine kinase. Plays a role in antigen receptor signaling for both clonal expansion and deletion in lymphoid cells. May also be involved in the regulation of gene expression. [UniProt].
Specifications
Specifications
| Antigen | HCLS1 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | PSH01-12 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | HCLS1 |
| Gene Accession No. | P14317 |
| Gene Alias | AW213261; CTTNL; HCLS1; hematopoietic cell specific Lyn substrate 1; hematopoietic cell-specific Lyn substrate 1; Hematopoietic lineage cell-specific protein; HS1; LckBP1; p75 |
| Show More |