missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ HBSS, calcium, magnesium, phenol red

Catalog No. A59772BA
Change view
Click to view available options
Quantity:
5 L
20 L
50 L
100 L
200 L
Format:
Drum
Sample
Tote
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
A59772BA 5 L Sample
A59772BB 20 L Tote
A59772BC 50 L Drum
A59772BD 100 L Drum
A59772BE 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. A59772BA Supplier Gibco™ Supplier No. A59772BA

Gibco™ Balanced Salt Solutions are sterile-filtered, packaged in bioprocess containers, and ready to order.

Gibco™ Balanced Salt Solutions are sterile-filtered, packaged in bioprocess containers, and ready to order. Commonly used for processing cells and flushing equipment, sourcing Gibco balanced salt solutions can help streamline your operation by reducing buffer preparation and allowing you to focus on additional critical steps in your biopharmaceutical processes. Discover new outsourcing efficiencies to reclaim time, space, and resources.

Product Features

  • Multiple formulations and concentrations available
  • Available in a range of pack sizes (5 L, 20 L, 50 L, 100 L, and 200 L)
  • Established products with set specifications are made to order
  • Product standardization enables shorter times from inquiry to quote and shipment, compared to custom
  • Readily accessible documentation for accelerated qualification

Benefits

  • Reduces time spent preparing buffers in-house
  • Mitigates space requirement for equipment storage
  • Streamlines in-house processes related to formulation, hydration, testing, and validation
  • Meets workflow needs across stages of development and manufacturing of biologicals

Specifications

Color Clear, colorless solution
Form Liquid
Product Type Hanks' Balanced Salt Solution (HBSS)
Sterility Sterile-filtered
Without Additives No Calcium, No Magnesium
Format Sample
Manufacturing Quality cGMP
Osmolality 265 to 295 mOsm/kg
pH 6.7 to 7.8
Quantity 5 L

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.