missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HBB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149846
Description
HBB Polyclonal Antibody for Western Blot, ICC/IF, IHC
The alpha (HBA) and beta (HBB) loci determine the structure of the 2 types of polypeptide chains in adult hemoglobin, Hb A. The normal adult hemoglobin tetramer consists of two alpha chains and two beta chains. Mutant beta globin causes sickle cell anemia. Absence of beta chain causes beta-zero-thalassemia. Reduced amounts of detectable beta globin causes beta-plus-thalassemia. The order of the genes in the beta-globin cluster is 5'-epsilon-gamma-G-gamma-A-delta-beta-3'.Specifications
| HBB | |
| Polyclonal | |
| Unconjugated | |
| HBB | |
| beta globin chain; beta-1-globin; Beta-globin; CD113t-C; Hbb; hemoglobin beta; Hemoglobin beta chain; hemoglobin beta chain complex; hemoglobin beta chain, major-form; Hemoglobin beta-1 chain; hemoglobin subunit beta; Hemoglobin subunit beta-1; hemoglobin, beta; III beta-1 globin; III beta-2 globin; LVV-hemorphin-7; Spinorphin | |
| Rabbit | |
| Protein A | |
| RUO | |
| 15127, 24440, 3043 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P02091, P68871 | |
| HBB | |
| Recombinant Hemoglobin Beta (HBb) (Met1-His147). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat, Chicken | |
| Antibody |