missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Hazardous Material Shipping Labels - Adhesive Vinyl
Hazardous Material Shipping Labels - Adhesive Vinyl: Permanent acrylic adhesive sticks to a wide variety of surfaces
Supplier: Brady™ 121471
Description
- Indoor/outdoor vinyl film withstands exposure to moisture, chemicals and handling.
- Up to 3 year average outdoor durability
Specifications
| Not Autoclavable | |
| Label | |
| Black on Yellow | |
| Vinyl | |
| Label | |
| English | |
| Rectangle | |
| 4 x 4 in. | |
| 149.86 mm |
| Globally Harmonized System of Classification and Labeling of Chemicals (GHS); NFPA 704; OSHA 1910.1200 | |
| Permanent Acrylic, Self-adhesive | |
| Label | |
| Single-side Printable | |
| ORGANIC PEROXIDE 5.2 | |
| 500/Roll | |
| 4 x 4 in. | |
| 4 in. | |
| 4 in. |