missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Hazardous Material Label, Legend: CONTAINS ASBESTOS…...CANCER AND LUNG DISEASE HAZARD
Description
- Abrasion and chemical resistant
- Does not include Write-On-Area
Specifications
Specifications
| Adhesive Type | Acrylic, Permanent |
| Color | Black, Red, White |
| Finish | Gloss |
| Material | Vinyl Film (B-7569) |
| Product Type | Hazardous Materials Label |
| Display Text | DANGER: CONTAINS ASBESTOS FIBERS AVOID CREATING DUST CANCER AND LUNG DISEASE HAZARD |
| Shape | Square |
| Size (English) | 4 x 4 in. |
| Size (Metric) | 10.16 x 10.16 cm (W x H) |
| Thickness (English) | 0.004 in. |