missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Hazardous Communication & Right-to-know Labels - Coated Paper
Coated Paper (B-235) features surface printed graphics on semi gloss paper label stock making it ideal for general label applications.
$52.97 - $144.08
Specifications
| Autoclavable | Not Autoclavable |
|---|---|
| Description | Label |
| Adhesive Type | Permanent Acrylic, Self-adhesive |
| Color | Black, Blue, Red, Yellow on White |
| Finish | Semi-gloss |
| Catalog Number | Mfr. No. | Dimensions (L x W) | Display Text | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Dimensions (L x W) | Display Text | Price | Quantity & Availability | ||||
|
19110380
|
Brady™
517LS |
0.875 x 2.875 in. | CHEMICAL: OWNER: DATE: |
Pack for $52.97
|
|
||||
|
19110329
|
Brady™
102844 |
1 x 3 in. | CHEMICAL NAME COMMON NAME MANUFACTURER DATE |
Each for $144.08
|
|
||||
|
19110430
|
Brady™
99421 |
7 x 7 in. | IDENTITY: CAS NO.: HEALTH FLAMMABILITY INSTABILITY PROTECTIVE EQUIPMENT |
Pack of 100 Each for $92.03
|
|
||||
|
19110428
|
Brady™
99188 |
4 x 5.875 in. | HEALTH FLAMMABILITY REACTIVITY PROTECTIVE EQUIPMENT |
Pack of 100 Each for $58.54
|
|
||||
Description
- B-235 Coated Paper with pressure sensitive acrylic adhesive from Brady™ adheres to most surfaces.
- Labels remove cleanly, leaving no residue on surface.
- Operating temperature of -40°F to +150°F (-40°C to +65.5°C).
- Recommended for indoor use.
Specifications
| Not Autoclavable | |
| Permanent Acrylic, Self-adhesive | |
| Semi-gloss | |
| Paper | |
| English | |
| 0.097 mm |
| Label | |
| Black, Blue, Red, Yellow on White | |
| Label | |
| Non-sterile | |
| 3.8 mil |