Learn More
New Pig Haz-Mat Pillows
Stop the spread of acids and bases.
Supplier: New Pig HR7015
Description
- Chemical-resistant — won't degrade or cause a dangerous reaction upon contact with corrosive spills
- Excellent for quickly diking and stopping the spread of chemical spills
- Absorb and retain most acids, bases and unknown liquids
- Can be incinerated after use to reduce waste
Specifications
| Pink | |
| 3.78 L | |
| Hazmat Absorbent Pillow | |
| 16 L x 17 W x 2 in. H | |
| 16 x 17 x 2 in. (40.6 x 43.18 x 5.08 cm) | |
| 40.6 cm | |
| 43.18 cm | |
| 5.08 cm | |
| 7.1 kg |
| 1 gal. | |
| Polypropylene | |
| Quickly diking and stopping the spread of chemical spills, absorbs acids, bases, corrosives | |
| 16 x 17 in. (40.6 x 43.18 cm) | |
| 16 in. | |
| 17 in. | |
| 2 in. | |
| 15.7 lb. |