missing translation for 'onlineSavingsMsg'
Learn More

Haws™ Test Kit

Catalog No. 18888334
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18888334 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18888334 Supplier Haws™ Supplier No. 9011

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Haws™ Test Kit is an ANSI compliance test kit.

Model 9011 is the new ANSI Emergency Shower and Eyewash Testing Kit. It includes a 5-gallon bucket with lid and marked line indicating the 2 gallon mark, a shower sock and 3-piece pole to direct water into bucket, eyewash gauge, tape measure, thermometer, ANSI checklist for testing compliance, and testing instructions. All items included fit directly into the bucket for ease of transportation.

Warranty

1 year from ship date (Limited)

Specifications

Certifications/Compliance ANSI Z358.1
Color Yellow
For Use With (Application) Emergency Shower
Height (English) 78 in.
Height (Metric) 1981 mm
Length (Metric) Handle 902 mm
Material Plastic
Product Type ANSI Shower and Eyewash Testing Kit