Learn More
Haws™ Freezeproof Emergency Station
Stays ready for emergency use in freezing temperatures
Supplier: Haws™ 8317CTFP
Includes:
Universal language emergency sign
Description
Freeze-protected 120 VAC cable heated combination shower and eye/face wash shall include the AXION MSR hydrodynamic designed green ABS plastic 10-5/8 in. (26.9 cm) showerhead with integral self-regulating flow control, an AXION MSR eye/face wash head.
- Perfect for extreme weather conditions dropping down to -30°F (-34.4°C)
- Universal design allows for top or bottom water supply
- Reducing installation time, entire unit is pre-built and fully water/pressure tested
- Solid stainless steel pipe stanchion and galvanized fittings make for one sturdy piece of corrosive-resistant equipment
- Rated Class 1, Div 2, Group B, C & D. Tempering code T6
- Certified by CSA to meet the ANSI Z358.1 Standard for Emergency Eyewash and Shower Equipment. Model is CSA electrical certified, TYPE 4. (Nema 4 equivalent)
Shower Head
- Shower head is equipped with 20 gpm (75.7L/min.) flow control
Eye/Facewash Heads
- Eye/facewash operates independently of shower
- ABS plastic Feather-Flo™ heads are angled to direct soft spray into the eyes and face
- Flip-top dust covers release automatically when water is turned on
Drench Hose
- Drench hose is operated independently by stay-open chrome-plated brass ball valve
Pipe and Fittings
- All valves and pipes are protected in an ultraviolet-resistant, Safety Green ABS plastic jacket
- Features removable polyethylene foam insulation and elastomeric seals, and a stainless-steel pull rod
- Piping stanchion is made of Type 304 Schedule 40 stainless steel
- Top and bottom supply, 11/4 in. (3.18cm) IPS
Order Info
Station operates on 120VAC 60Hz, single phase. A Class 1, Division 1 model is available on request; contact your Fisher Safety Customer Service Representative.
Certifications
CSA certified to ANSI™ Z358.1
Specifications
| ANSI Z358.1 | |
| Emergency use in Freezing Temperatures, for Weather Conditions dropping down to -34.4°C | |
| Freezeproof Emergency Station |
| 20 gpm | |
| ABS Plastic feather-Flo™ eye/facewash head, self regulating heat-trace cable, Thermostat guard, Stainless Steel pipe stanchion, drench hose |