missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Haws™ Deck Mount Eye/Face Wash
Haws™ Deck Mount Eye/Face Wash features an AXION™ MSR eye/face wash and body spray head, with 8-foot green hose
Supplier: Haws™ 8904
Description
- Eyewash streams provide Zero Vertical Velocity™ stream engineering with even laminar flow for enhanced comfort, stability and effectiveness
- Dust cover allows for protection from debris when not in use
- Easily detected and maintained, emergency equipment is equipped with safety sign and test card to record weekly testing
- 8 foot (243.8cm) green hose provides an extended coverage area to further optimize the irrigation process
- In order to comply with state codes, the use of a vacuum breaker is required with all hand held eye/face wash body spray units
Warranty
1 year from ship date (Limited)
Specifications
| Squeeze Handle | |
| Green, Yellow, Silver | |
| 104 in. | |
| 1.27 cm | |
| Countertop | |
| 5 in. |
| ANSI Z358.1 | |
| 3.7 gpm | |
| 264.16 cm | |
| Polished Chrome, Plastic | |
| 250 psi | |
| 12.7 cm |