missing translation for 'onlineSavingsMsg'
Learn More

Haws™ AXION MSR™ Swing-Away Eye/Face Wash

Catalog No. 171602
Change view
Click to view available options
Flow Rate:
1.2 gpm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate
171602 1.2 gpm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 171602 Supplier Haws™ Supplier No. 7612

missing translation for 'validMainframeMsg'

Includes

Universal language sign and waterproof test tag.

AXION MSR™ countertop swing-away eye/face wash

  • Designed to be mounted on right side of sink
  • Dust cover offers protection from debris when not in use
  • Requires minimal maintenance and is equipped with test card to record weekly testing

Specifications

Certifications/Compliance ANSI Z358.1
Flow Rate 1.2 gpm
For Use With (Application) Sweeping Contaminants away from the Vulnerable Nasal Cavity
Includes Stainless Steel eye/facewash head, green PE Plastic dust cover, Chrome Plated Brass flow control assembly, universal language sign, Waterproof test tags
Mounting Countertop
Pressure 30 to 90 psi
Product Type Laboratory Eyewash