missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| HATU | |
| 186°C | |
| C10H15F6N6OP | |
| 25 g | |
| JNWBBCNCSMBKNE-UHFFFAOYSA-N | |
| [(dimethylamino)({3H-[1,2,3]triazolo[4,5-b]pyridin-3-yloxy})methylidene]dimethylazanium; hexafluoro-λâµ-phosphanuide | |
| 9886157 | |
| ≥98.0% (HPLC) |
| 148893-10-1 | |
| White-Yellow | |
| MFCD00274639 | |
| hatu, 2-7-aza-1h-benzotriazole-1-yl-1,1,3,3-tetramethyluronium hexafluorophosphate, o-7-azabenzotriazol-1-yl-n,n,n',n'-tetramethyluronium hexafluorophosphate, o-7-azabenzotriazol-1-yl-n,n,n,n-tetramethyl uronium hexafluorophosphate, 2-3h-1,2,3 triazolo 4,5-b pyridin-3-yl-1,1,3,3-tetramethyluronium hexafluorophosphate v, dimethylamino triazolo 4,5-b pyridin-3-yloxy methylene-dimethyl-ammonium hexafluorophosphate, 2-7-aza-1h-benzotriazole-1-yl-1,1,3,3-tetramethyl uronium hexafluorophosphate, 2-3h-1,2,3 triazolo 4,5-b pyridin-3-yl-1,1,3,3-tetramethylisouronium hexafluorophosphate v, n-dimethylamino 3h-1,2,3 triazolo 4,5-b pyridin-3-yloxy methylene-n-methylmethanaminium hexafluorophosphate, n-dimethylamino-1h-1,2,3-triazolo-4,5-b pyridin-1-ylmethylene-n-methylmethanaminium hexafluorophosphate n-oxide | |
| F[P-](F)(F)(F)(F)F.CN(C)C(ON1N=NC2=CC=CN=C12)=[N+](C)C | |
| 380.24 | |
| 380.24 | |
| Crystalline Powder |
Chemical Identifiers
| 148893-10-1 | |
| 380.24 | |
| JNWBBCNCSMBKNE-UHFFFAOYSA-N | |
| 9886157 | |
| F[P-](F)(F)(F)(F)F.CN(C)C(ON1N=NC2=CC=CN=C12)=[N+](C)C |
| C10H15F6N6OP | |
| MFCD00274639 | |
| hatu, 2-7-aza-1h-benzotriazole-1-yl-1,1,3,3-tetramethyluronium hexafluorophosphate, o-7-azabenzotriazol-1-yl-n,n,n',n'-tetramethyluronium hexafluorophosphate, o-7-azabenzotriazol-1-yl-n,n,n,n-tetramethyl uronium hexafluorophosphate, 2-3h-1,2,3 triazolo 4,5-b pyridin-3-yl-1,1,3,3-tetramethyluronium hexafluorophosphate v, dimethylamino triazolo 4,5-b pyridin-3-yloxy methylene-dimethyl-ammonium hexafluorophosphate, 2-7-aza-1h-benzotriazole-1-yl-1,1,3,3-tetramethyl uronium hexafluorophosphate, 2-3h-1,2,3 triazolo 4,5-b pyridin-3-yl-1,1,3,3-tetramethylisouronium hexafluorophosphate v, n-dimethylamino 3h-1,2,3 triazolo 4,5-b pyridin-3-yloxy methylene-n-methylmethanaminium hexafluorophosphate, n-dimethylamino-1h-1,2,3-triazolo-4,5-b pyridin-1-ylmethylene-n-methylmethanaminium hexafluorophosphate n-oxide | |
| [(dimethylamino)({3H-[1,2,3]triazolo[4,5-b]pyridin-3-yloxy})methylidene]dimethylazanium; hexafluoro-λâµ-phosphanuide |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator