accessibility menu, dialog, popup

UserName

CAS RN 148893-10-1

IUPAC Name: [(dimethylamino)({3H-[1,2,3]triazolo[4,5-b]pyridin-3-yloxy})methylidene]dimethylazanium; hexafluoro-λ⁵-phosphanuide
Synonyms: HATU
Molecular Weight (g/mol): 380.24
Molecular Formula: C10H15F6N6OP
InChi Key: JNWBBCNCSMBKNE-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.CN(C)C(ON1N=NC2=CC=CN=C12)=[N+](C)C
Molecular Weight (g/mol) 
  • (1)
  • (5)
Quantity 
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)