missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Harmine Hydrochloride 98.0+%, TCI America™
$37.29 - $219.95
Chemical Identifiers
| CAS | 343-27-1 |
|---|---|
| Molecular Formula | C13H13ClN2O |
| Molecular Weight (g/mol) | 248.71 |
| MDL Number | MFCD00012641 |
| InChI Key | VNPLYCKZIUTKJM-UHFFFAOYSA-N |
| Synonym | Banisterine Hydrochloride |
| PubChem CID | 5359389 |
| IUPAC Name | hydrogen 7-methoxy-1-methyl-9H-pyrido[3,4-b]indole chloride |
| SMILES | [H+].[Cl-].COC1=CC2=C(C=C1)C1=C(N2)C(C)=NC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
H0002100MG
|
TCI America
H0002100MG |
100 mg |
Each for $37.29
|
|
|||||
|
H00021G
|
TCI America
H00021G |
1 g |
Each for $219.95
|
|
|||||
Chemical Identifiers
| 343-27-1 | |
| 248.71 | |
| VNPLYCKZIUTKJM-UHFFFAOYSA-N | |
| 5359389 | |
| [H+].[Cl-].COC1=CC2=C(C=C1)C1=C(N2)C(C)=NC=C1 |
| C13H13ClN2O | |
| MFCD00012641 | |
| Banisterine Hydrochloride | |
| hydrogen 7-methoxy-1-methyl-9H-pyrido[3,4-b]indole chloride |
Specifications
| 343-27-1 | |
| C13H13ClN2O | |
| 100 mg | |
| Banisterine Hydrochloride | |
| [H+].[Cl-].COC1=CC2=C(C=C1)C1=C(N2)C(C)=NC=C1 | |
| 248.71 | |
| 248.71 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00012641 | |
| 1544 | |
| VNPLYCKZIUTKJM-UHFFFAOYSA-N | |
| hydrogen 7-methoxy-1-methyl-9H-pyrido[3,4-b]indole chloride | |
| 5359389 | |
| ≥98.0% (HPLC,T) | |
| Harmine Hydrochloride |
Safety and Handling
RTECSNumber : MG9450000
TSCA : No